missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brij™-35 Surfact-Amps™ Detergent Solution

Catalog No. PI85118
Change view
Click to view available options
Quantity:
50 mL
500 mL
6 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI85118 500 mL
PI28316 6 x 10 mL
PI85117 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI85118 Supplier Thermo Scientific™ Supplier No. 85118
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.

Thermo Scientific™ Brij-35 Detergent Solutions are stabilized 10% and 30% solutions of Brij-35 detergent for use in various protein methods.

Additional Information and Online Ordering

Brij-35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or as a surfactant in various HPLC applications. The detergent is offered in two forms: a Thermo Scientific Surfact-Amps™ Detergent Solution (high-purity, 10% solution) and a standard grade 30% solution.

Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neet detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

  • Brij-35 – a nonionic detergent for use in various protein methods

  • Accurate – 10% and 30% detergent solutions in ultrapure water

  • Easy-to-use – solutions are simple to dispense and dilute for use

Detergent Properties:

  • Molecular Weight: 1225g
  • Detergent Class: Nonionic
  • Aggregation Number: 40
  • Micelle Molecular Weight: 49,000g
  • Critical Micelle Concentration (CMC): 0.09mM (0.011%, w/v)
  • Cloud Point: >100°C
  • Dialyzable: No

Surfact-Amps Detergent Solution (Part No. 28316):

  • Accurate – precise 10% detergent solution in ultrapure water

  • Easy-to-use – solution is simple to dispense and dilute for use

  • Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

  • Stable – packed in glass ampules under inert nitrogen gas

Specifications

Content And Storage Store at Room Temperature
Form Liquid
Product Line Brij, Surfact-Amps
Reagent Type Detergent Solution
Quantity 500 mL
What is the Brij 35 detergent concentration in the 1X Zymogram Developing buffer?

The Brij 35 detergent concentration for Zymogram Developing buffer should be 0.02% (w/v).

For Research Use Only. Not for use in diagnostic procedures.