missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brij™-35, 30% Solution

Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.

Supplier:  Thermo Scientific™ LSG20150

Numéro de catalogue. PI20150


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific formulation of Brij 35 is a stabilized 30% (w/w) solution of Brij-35 detergent for use in various protein methods. Brij 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications.

Properties of Brij 35 Detergent:

• Molecular Weight: 1225 g
• Detergent Class: Non-ionic
• Aggregation Number: 40
• Micelle Molecular Weight: 49,000 g
• Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
• Cloud Point: >100°C
• Dialyzable: No

Features of this Thermo Scientific detergent solution:

• A nonionic detergent for use in various protein methods
Accurate—30% detergent solution in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use

Specifications of this Thermo Scientific Brij 35 30% solution:

• Visual: Clear, colorless viscous liquid, free of foreign material.
• Concentration: 27.0 to 30.0%

Spécifications

Spécifications

Brij-35, 30% Solution
Store at Room Temperature
Brij, Surfact-Amps
950 mL
Detergent
Liquid
Detergent Solution
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.