missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brij 35, 30% (w/w) Aqueous Wetting Agent, Ricca Chemical
$160.61 - $286.60
Specifications
| Chemical Name or Material | Brij 35 |
|---|---|
| CAS | 7732-18-5 , 9002-92-0 |
| Packaging | Natural Poly Bottle |
| Grade | Laboratory |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
114516
|
Ricca Chemical Company
114516 |
500 mL |
Each for $160.61
|
|
|||||
|
1145-32
|
Ricca Chemical Company
114532 |
1 L |
Each for $286.60
|
|
|||||
Specifications
| Brij 35 | |
| 7732-18-5,9002-92-0 | |
| Natural Poly Bottle | |
| Laboratory |