missing translation for 'onlineSavingsMsg'
Learn More

Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35

CAS: 9002-92-0 | C58H118O24

Supplier:  Thermo Scientific Chemicals 329580025

 View more versions of this product

Catalog No. AC329580025

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

Colorless
Jelly Substance at 25°C
Glass bottle
C58H118O24
MFCD00080891
15, 7675
1199.56
Brij™ 35
9002-92-0 , 7732-18-5
1.0300g/mL
1.03
C12H25(OCH2CH2)23OH
06,70
Solubility in water: soluble.
29 to 31%

Specifications

Brij™ 35
9002-92-0 , 7732-18-5
1.0300g/mL
Glass bottle
C58H118O24
MFCD00080891
15, 7675
1199.56
Colorless
Jelly Substance at 25°C
2.5 L
1.03
C12H25(OCH2CH2)23OH
06,70
Solubility in water: soluble.
29 to 31%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only