accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Facility Safety and Maintenance
Cleaning Supplies and Equipment
Cleaners and Disinfectants
Laboratory Surfactants and Wetting Agents
LC117921
Print
Brij 35, 30% (w/v), LabChem™
Supplier:
LabChem
LC117921
Catalog No.
LC117921
Sign In
or
Register
to check your price and availability.
$125.39
/
Each
Quantity
Only null left
Explore available promotions
Add to cart
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Promotions
Description
Description
Specifications
Specifications
Chemical Name or Material
Brij 35
CAS
9002-92-0
,
7732-18-5
CAS Max %
70,30
Quantity
500 mL
Packaging
Poly Bottle
Solubility Information
Soluble in water
Color
Colorless
Physical Form
Liquid
Decomposition Information
Fume; Carbon monoxide; Carbon dioxide
Identification
Passes Test
Linear Formula
(C
2
H
4
O)
n
C
1
2
H
2
6
O
Percent Purity
30% (w/v)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions