missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Bright-Glo™ Luciferase Assay System
Bright-Glo™ Luciferase Assay System is designed for continuous-process systems. The robust, homogeneous assay chemistry gives high assay sensitivity and has a 30-minute signal half life.
$287.24 - $14187.92
Specifications
| Content And Storage | -30°C to -10°C |
|---|---|
| For Use With (Application) | Assays and biosensors that monitor cell-signaling pathways, bioluminescence resonance energy transfer (BRET) to monitor Protein-Protein interactions |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR-E2620
|
Promega
E2620 |
100 mL (1 Pk.) |
Each for $1,712.88
|
|
|||||
PR-E2610
|
Promega
E2610 |
10 mL (1 Pk.) |
Each for $287.24
|
|
|||||
PR-E2650
|
Promega
E2650 |
10 x 100 mL (1 Pk.) |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Sensitive Reporter Assay for Continuous Processing on Robotic Systems
- Sensitive, bright signal with 30-minute half-life
- No need to remove culture medium or wash cells prior to adding assay reagent
- Less sensitive to mixing conditions, sample evaporation and pipetting errors
- U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents and patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
- Certain applications of this product may require licenses from others.
Specifications
| -30°C to -10°C | |
| Assays and biosensors that monitor cell-signaling pathways, bioluminescence resonance energy transfer (BRET) to monitor Protein-Protein interactions |