missing translation for 'onlineSavingsMsg'
Learn More

Promega Bright-Glo™ Luciferase Assay System

Catalog No. PRE2620
Encompass
Change view
Click to view available options
Quantity:
10 mL (1 Pk.)
10 x 100 mL (1 Pk.)
100 mL (1 Pk.)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE2620 100 mL (1 Pk.)
PRE2610 10 mL (1 Pk.)
PRE2650 10 x 100 mL (1 Pk.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRE2620 Supplier Promega Supplier No. E2620

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Bright-Glo Luciferase Assay Substrate; Bright-Glo Luciferase Assay Buffer

Bright-Glo™ Luciferase Assay System is designed for continuous-process systems. The robust, homogeneous assay chemistry gives high assay sensitivity and has a 30-minute signal half life.

  • Sensitive Reporter Assay for Continuous Processing on Robotic Systems
  • Sensitive, bright signal with 30-minute half-life
  • No need to remove culture medium or wash cells prior to adding assay reagent
  • Less sensitive to mixing conditions, sample evaporation and pipetting errors

  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  • Certain applications of this product may require licenses from others.

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Assays and biosensors that monitor cell-signaling pathways, bioluminescence resonance energy transfer (BRET) to monitor Protein-Protein interactions
Quantity 100 mL (1 Pk.)