Learn More
Invitrogen™ eBioscience™ Brefeldin A Solution (1000X)
Catalog No. 501129757
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
501129757 1 mL
1 options
Catalog No. 501129757 Supplier Invitrogen™ Supplier No. 00450651
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Addition of Brefeldin A during the last hours of in vitro activation of cells results in enhanced detection of intracellular cytokines.
  • Brefeldin A is an inhibitor of intracellular protein transport
  • Incubation of cells in culture with Brefeldin A leads to blockade of protein transport to the Golgi complex (GC) and accumulation of proteins in the endoplasmic reticulum (ER)
  • Brefeldin A is effective for enhanced detection of a majority of mouse and human intracellular cytokines; however, it is advised that the investigators evaluate the use and efficacy of this reagent as well as other protein transport inhibitors such as Monensin in their specific assay system
Specifications
Quantity 1 mL
Product Line eBioscience
Physical Form Liquid
Product Type Inhibitor
For Research Use Only. Not for use in diagnostic procedures.