missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ 384-Well Microplates
Non-treated microplates for diverse applications
$623.63 - $2940.74
Specifications
| Barcode | No Barcode |
|---|---|
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Material | Polystyrene |
| Catalog Number | Mfr. No. | Color | Volume (Metric) Well | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Volume (Metric) Well | Price | Quantity | ||||
|
14-386-058
|
N/A | Clear | 0.1 mL |
Pack of 50 Each for $623.63
|
|
||||
|
14-386-013
|
N/A | White | 0.1 mL |
Pack of 50 Each for $1,085.37
|
|
||||
|
14-386-014
|
N/A | Black | 0.1 mL |
Pack of 50 Each for $1,158.22
|
|
||||
|
14-386-019
|
N/A | White, Clear Bottom | 0.12 mL |
Pack of 50 Each for $2,940.74
|
|
||||
|
14-386-020
|
N/A | Black, Clear Bottom | 0.12 mL |
Pack of 50 Each for $2,940.74
|
|
||||
Description
- Non-treated PS surface
- Medium binding, non-sterile
- Standard plate for most applications
- Free from endotoxins, DNase, DNA, RNase; noncytoxic
Specifications
| No Barcode | |
| Without Lid | |
| Polystyrene | |
| Non-sterile | |
| Flat Bottom (F-well) | |
| 50/Cs. |
| Standard | |
| 16 x 24 | |
| 384 | |
| Non-treated | |
| For homogenous assays, screenings, storage |