Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ 384-Well Microplates
Non-treated microplates for diverse applications
 View more versions of this product
Catalog No. 14-386-058
Only null left
Explore available promotions
Description
Description
  • Non-treated PS surface
  • Medium binding, non-sterile
  • Standard plate for most applications
  • Free from endotoxins, DNase, DNA, RNase; noncytoxic
Specifications
Specifications
No Barcode
Without Lid
Clear
0.1 mL
Non-sterile
Flat Bottom (F-well)
50/Cs.
Standard
16 x 24
Polystyrene
384
Non-treated
For homogenous assays, screenings, storage
Product Suggestions
Product Suggestions