Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ 384-Well Microplates
Non-treated microplates for diverse applications
Description
- Non-treated PS surface
- Medium binding, non-sterile
- Standard plate for most applications
- Free from endotoxins, DNase, DNA, RNase; noncytoxic
Specifications
| No Barcode | |
| Without Lid | |
| Black, Clear Bottom | |
| 0.12 mL | |
| Non-sterile | |
| Flat Bottom (F-well) | |
| 50/Cs. |
| Standard | |
| 16 x 24 | |
| Polystyrene | |
| 384 | |
| Non-treated | |
| For homogenous assays, screenings, storage |