Learn More
BRANDTECH™ BRAND™ Titrette™ Digital Bottle-Top Burettes
GREENER_CHOICE
Provides accuracy and precision within the tolerances of Class A glass burettes. Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
$5,561.32 - $8,131.46
Specifications
Product Type Digital Bottletop Dispenser
Angle 90°
Autoclavable Not Autoclavable
Certifications/Compliance DIN EN ISO 385, ASTM E287
Color Gray
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Interface Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Interface Price Quantity & Availability  
14379342
GREENER_CHOICE
View Documents Promotion Details
BRANDTECH™ BRAND™
4760151
25 mL Without Interface
Each for $5,889.43
Only null left
14379343
GREENER_CHOICE
View Documents Promotion Details
BRANDTECH™ BRAND™
4760161
50 mL Without Interface
Each for $5,889.43
Only null left
14379340
GREENER_CHOICE
View Documents Promotion Details
BRANDTECH™ BRAND™
4760251
25 mL RS232 Interface
Each for $8,131.46
Only null left
14379341
GREENER_CHOICE
View Documents Promotion Details
BRANDTECH™ BRAND™
4760261
50 mL RS232 Interface
Each for $7,678.43
Only null left
13690100
GREENER_CHOICE
View Documents Promotion Details
BRANDTECH™ BRAND™
4760141
10 mL RS232 Interface
Each for $5,561.32
Only null left
13690101
GREENER_CHOICE
View Documents Promotion Details
BRANDTECH™ BRAND™
4760241
10 mL RS232 Interface
Each for $7,678.43
Only null left
Description
Description
Provides accuracy and precision within the tolerances of Class A glass burettes. BrandTech™ BRAND™ Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
The Titrette™ bottle-top burette provides accuracy and precision within the tolerances of Class A glass burettes. Suitable for most titrants in concentrations up to 1 mol/L, it is ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more. The Titrette will enable you to perform routine titrations quickly, accurately, and with ease.
  • Smooth operation for exceptionally sensitive drop-wise titrations
  • Avoids the risk of spills from poured transfers
  • Eliminates meniscus reading errors
  • Switch automatically between filling and titrating
  • Meets tolerances for Class A glass burettes per ISO and ASTM standards
  • Optional RS-232 interface available for direct data transmission to a PC
  • Use for a variety of applications including general chemistry, water treatment, food/beverage analysis, industrial titrations, and environmental work in the lab or field
Order Info
For burettes with Calibration Certificate, contact Customer Service. Add DKD at the end of the BRANDTECH part number. No returns accepted.
Specifications
Specifications
Digital Bottletop Dispenser
Not Autoclavable
Gray
Digital
15°C to 40°C
1.5 V
90°
DIN EN ISO 385, ASTM E287
Glass
59°F to 104°F
Up to 500 mm 2/s