Learn More
BrandTech™ BRAND™ BLAUBRAND™ Glass Volumetric Flasks, Class A, USP, Certified
Indispensable for preparing dilutions and standard solutions
$229.31 - $854.84
Specifications
| Calibration | To Contain |
|---|---|
| Certifications/Compliance | DIN EN ISO 1042, USP batch certificate |
| Closure Material | Glass |
| Material | Borosilicate Glass (3.3) |
| Product Type | Volumetric Flask |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Color | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Color | Price | Quantity | |||
|
13-690-127
|
BrandTech™ BRAND™
36968 |
5 mL | NS 10/19 | Clear |
Pack of 2 Each for $229.31
|
|
|||
|
13-690-128
|
BrandTech™ BRAND™
36973 |
10 mL | NS 10/19 | Clear |
Pack of 2 Each for $229.31
|
|
|||
|
13-690-130
|
BrandTech™ BRAND™
36977 |
25 mL | NS 10/19 | Clear |
Pack of 2 Each for $229.31
|
|
|||
|
13-690-131
|
BrandTech™ BRAND™
36978 |
50 mL | NS 12/21 | Clear |
Pack of 2 Each for $245.25
|
|
|||
|
13-690-132
|
BrandTech™ BRAND™
36979 |
100 mL | NS 14/23 | Clear |
Pack of 2 Each for $268.19
|
|
|||
|
13-690-133
|
BrandTech™ BRAND™
36980 |
200 mL | NS 14/23 | Clear |
Pack of 2 Each for $354.19
|
|
|||
|
13-690-134
|
BrandTech™ BRAND™
36981 |
250 mL | NS 14/23 | Clear |
Pack of 2 Each for $371.38
|
|
|||
|
13-690-135
|
BrandTech™ BRAND™
36982 |
500 mL | NS 19/26 | Clear |
Pack of 2 Each for $448.44
|
|
|||
|
13-690-136
|
BrandTech™ BRAND™
36983 |
1000 mL | NS 24/29 | Clear |
Each for $349.72
|
|
|||
|
13-690-137
|
BrandTech™ BRAND™
36984 |
2000 mL | NS 29/32 | Clear |
Each for $466.28
|
|
|||
|
13-690-138
|
BrandTech™ BRAND™
37461 |
5 mL | NS 10/19 | Amber |
Pack of 2 Each for $487.31
|
|
|||
|
13-690-139
|
BrandTech™ BRAND™
37462 |
10 mL | NS 10/19 | Amber |
Pack of 2 Each for $487.31
|
|
|||
|
13-690-140
|
BrandTech™ BRAND™
37463 |
20 mL | NS 10/19 | Amber |
Pack of 2 Each for $487.31
|
|
|||
|
13-690-141
|
BrandTech™ BRAND™
37464 |
25 mL | NS 10/19 | Amber |
Pack of 2 Each for $487.31
|
|
|||
|
13-690-142
|
BrandTech™ BRAND™
37465 |
50 mL | NS 12/21 | Amber |
Pack of 2 Each for $504.50
|
|
|||
|
13-690-143
|
BrandTech™ BRAND™
37467 |
100 mL | NS 14/23 | Amber |
Pack of 2 Each for $504.50
|
|
|||
|
13-690-144
|
BrandTech™ BRAND™
37468 |
200 mL | NS 14/23 | Amber |
Pack of 2 Each for $641.47
|
|
|||
|
13-690-145
|
BrandTech™ BRAND™
37469 |
250 mL | NS 14/23 | Amber |
Pack of 2 Each for $700.72
|
|
|||
|
13-690-146
|
BrandTech™ BRAND™
37470 |
500 mL | NS 19/26 | Amber |
Pack of 2 Each for $854.84
|
|
|||
|
13-690-147
|
BrandTech™ BRAND™
37471 |
1000 mL | NS 24/29 | Amber |
Each for $623.63
|
|
|||
|
13-690-129
|
BrandTech™ BRAND™
36975 |
20 mL | NS 10/19 | Clear |
Pack of 2 Each for $229.31
|
|
|||
Description
- Manufactured from Borosilicate 3.3 glass, known for its low expansion coefficient and excellent chemical resistance
- Available in transparent or amber glass
- Glass stopper with drip-tip included
- Featuring a full range of ISO size options, from 5 mL to 2000 mL
- Calibrated 'To Contain'
- Each unit is supplied with a batch number and a USP batch certificate in its original packaging for traceability
Specifications
| To Contain | |
| Glass | |
| Volumetric Flask | |
| Reusable | |
| Narrow | |
| A |
| DIN EN ISO 1042, USP batch certificate | |
| Borosilicate Glass (3.3) | |
| Autoclavable | |
| No | |
| Non-sterile | |
| Round |