Learn More
BrandTech™ BRAND™ BLAUBRAND™ Glass Volumetric Flasks, Class A, USP, Certified
Indispensable for preparing dilutions and standard solutions
Supplier:  BrandTech™ BRAND™ 37462
 View more versions of this product
Catalog No. 13-690-139
Only null left
Explore available promotions
Description
Description
  • Manufactured from Borosilicate 3.3 glass, known for its low expansion coefficient and excellent chemical resistance
  • Available in transparent or amber glass
  • Glass stopper with drip-tip included
  • Featuring a full range of ISO size options, from 5 mL to 2000 mL
  • Calibrated 'To Contain'
  • Each unit is supplied with a batch number and a USP batch certificate in its original packaging for traceability
Specifications
Specifications
To Contain
Glass
10 mL
±0.02 mL USP
Volumetric Flask
Reusable
Narrow
A
NS 10/19
DIN EN ISO 1042, USP batch certificate
Amber
Borosilicate Glass (3.3)
2 Pieces
Autoclavable
No
Non-sterile
Round
Product Suggestions
Product Suggestions