Learn More
Brady™ ToughStripe™ Labels with Overlaminate
Create custom-printed floor labels quickly and easily
Supplier:  Brady™ 142167
 View more versions of this product
Catalog No. 19150888
Only null left
Includes:
  • 1 roll of B-483 polyester
  • 1 roll of B-634 polyester overlaminate
Description
Description
Easy Three-Step Process
  • Print label
  • Position on floor
  • Apply overlaminate
Specifications
Chemical Identifiers
RoHS 2002/95/EC
None
Polyester
Thermal Transfer
Non-sterile
GlobalMark™ 2 and BMP™ 71 label printer
1
1 Kit
-18°C to +54°C
0.203 mm
Rubber, Permanent
Gloss
Single-side Printable
Label
For high adhesion to textured metals and low surface energy Plastics
1 Roll of tape and 1 roll of over laminate
-18°C
0°F to 130°F
0.004 in.
Specifications
RoHS 2002/95/EC
None
Gloss
Polyester
Thermal Transfer
Non-sterile
GlobalMark™ 2 and BMP™ 71 label printer
1
2 Rolls/Kit
0°F to 130°F
0.004 in.
100 ft. x 4 in. (30.48 m x 10.2 cm)
30.48 m
10.2 cm
Rubber, Permanent
Red, Clear
Tape
Single-side Printable
Label
For high adhesion to textured metals and low surface energy Plastics
1 Roll of tape and 1 roll of over laminate
-18°C
1 Kit
-18°C to +54°C
0.203 mm
100 ft.
4 in.
Product Suggestions
Product Suggestions