Learn More
Brady™ ToughStripe™ Floor Marking Corner Marks
Durable tape perfect for marking skid and pallet storage areas, as well as other designated areas
Supplier:  Brady™ 104454
 View more versions of this product
Catalog No. 19150784
Only null left
Description
Description
Durability
  • Withstands forklift traffic without tearing or lifting
  • Low-profile design minimizes tears and scratches from skids, pallet jacks, etc.
  • Made with rugged polyester and aggressive adhesive
Painless Removal
  • Does not chip and flake when being removed - comes off in one piece
  • Clean removal without any unsightly adhesive residue or damage to the floor
High Visibility
  • High-gloss surface shines like new paint
  • Low profile minimizes debris buildup along the edges that occurs with thicker (extruded) floor marks
  • Surface resists marks and smudges and cleans up like new
  • Available in a variety of high-impact colors
Specifications
Chemical Identifiers
ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
Gloss
Polyester
None
Non-sterile
115 Gardner Units
20 Pieces/Pack
L-shaped
-18°C to +54°C
0.201 mm
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
None
Tape
Single-side Printable
Label Tape
For floor corner mark, Used to mark aisles, passageways, storage locations and point out safety equipment and exits
-18°C
20/Pk.
0°F to 130°F
0.008 in.
Specifications
ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
Red
Tape
Single-side Printable
Label Tape
For floor corner mark, Used to mark aisles, passageways, storage locations and point out safety equipment and exits
-18°C
20/Pk.
0°F to 130°F
0.008 in.
254 x 101.60 mm
25.4 cm
101.60 mm
254.00 mm
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
None
Gloss
Polyester
None
Non-sterile
115 Gardner Units
20 Pieces/Pack
L-shaped
-18°C to +54°C
0.201 mm
10 in.
4 in.
10 in.
TS-101.60-514-COR-RD-20
Product Suggestions
Product Suggestions