missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Safety First: Wear Face Shields, Rubber Gloves... Signs
Identify equipment, rooms and materials which contain or are contaminated with hazardous agents
$34.50 - $72.97
Specifications
| Quantity | 1 Ea. |
|---|---|
| Mounting Type | Surface Mount |
| Min. Temperature (English) | -40°F |
| Displayed Text | Safety First Wear Face Shields, Rubber Gloves And Aprons When Working With Acids |
| Product Type | Protective Wear Sign |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Temperature Range (English) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Temperature Range (English) | Price | Quantity | |||
|
19-016962
|
Brady™
74464 |
20 x 14 in. | Fiberglass | -40°F to 193°F |
Each for $72.97
|
|
|||
|
19-036341
|
Brady™
84478 |
14 x 10 in. | Polyester | -40°F to 230°F |
Each for $34.50
|
|
|||
Description
- Sign reads “SAFETY FIRST - WEAR FACE SHIELDS, RUBBER GLOVES AND APRONS WHEN WORKING WITH ACIDS”
- Green/black on white
- Available in different sizes and materials
Specifications
| 1 Ea. | |
| -40°F | |
| Protective Wear Sign |
| Surface Mount | |
| Safety First Wear Face Shields, Rubber Gloves And Aprons When Working With Acids | |
| White |