missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Safety First: Wear Face Shields, Rubber Gloves... Signs

Catalog No. 19016962
Change view
Click to view available options
Dimensions (L x W):
14 x 10 in.
20 x 14 in.
Material:
Fiberglass
Polyester
Temperature Range (English):
-40°F to 193°F
-40°F to 230°F
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material Temperature Range (English)
19016962 20 x 14 in. Fiberglass -40°F to 193°F
19036341 14 x 10 in. Polyester -40°F to 230°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19016962 Supplier Brady™ Supplier No. 74464

Identify equipment, rooms and materials which contain or are contaminated with hazardous agents

  • Sign reads “SAFETY FIRST - WEAR FACE SHIELDS, RUBBER GLOVES AND APRONS WHEN WORKING WITH ACIDS”
  • Green/black on white
  • Available in different sizes and materials

Specifications

Certifications/Compliance ASTM D 1593, D 638, D 790, D 646, D 635
Material Fiberglass
Quantity 1 Ea.
Tensile Strength 9000 psi
Description Premium Fiberglass
Mounting Type Surface Mount
For Use With (Application) Industrial, utility, commercial and institutional environments for both indoor and outdoor use
Height (English) 14 in.
Height (Metric) 35.5 cm
Max. Temperature (English) 193°F
Min. Temperature (English) -40°F
Thickness 0.1 in.
Width (English) 20 in.
Width (Metric) 50.8 cm
Temperature Range (Metric) -40°C to 90°C
Temperature Range (English) -40°F to 193°F
Dimensions (L x W) 20 x 14 in.
Displayed Text Safety First Wear Face Shields, Rubber Gloves And Aprons When Working With Acids
Product Type Protective Wear Sign
Color White
Show More Show Less