missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Safety First: Wear Face Shields, Rubber Gloves... Signs

Catalog No. 19036341
Change view
Click to view available options
Dimensions (L x W):
14 x 10 in.
20 x 14 in.
Material:
Fiberglass
Polyester
Temperature Range (English):
-40°F to 193°F
-40°F to 230°F
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material Temperature Range (English)
19036341 14 x 10 in. Polyester -40°F to 230°F
19016962 20 x 14 in. Fiberglass -40°F to 193°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19036341 Supplier Brady™ Supplier No. 84478

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify equipment, rooms and materials which contain or are contaminated with hazardous agents

  • Sign reads “SAFETY FIRST - WEAR FACE SHIELDS, RUBBER GLOVES AND APRONS WHEN WORKING WITH ACIDS”
  • Green/black on white
  • Available in different sizes and materials

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145, PSTC-33,
Material Polyester
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
Height (English) 10 in.
Height (Metric) 25.4 cm
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.006 to 0.01 in.
Width (English) 14 in.
Width (Metric) 35.5 cm
Temperature Range (Metric) -40°C to 110°C
Shape Rectangular
Temperature Range (English) -40°F to 230°F
Languages English
Dimensions (L x W) 14 x 10 in.
Displayed Text Safety First Wear Face Shields, Rubber Gloves And Aprons When Working With Acids
Product Type Protective Wear Sign
Color White
Show More Show Less