missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads
Color-coded absorbents for safe clean-up of hazardous fluids. Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads are surfactant treated polypropylene pads used on wide range of chemicals.
Supplier: Brady™ BRDCH200
Description
- Benefits of sign and an absorbent in one
- Highly absorbent for all-purpose industrial applications
- Ideal for areas where the risk of slips, trips and falls are greatest
- Chemically inert, do not react with aggressive fluids
- Perforations provide cost control and reduce waste
- Heavy weight - specifically designed for continuous leaks and larger spills
- Light weight - specifically designed for smaller leaks, drips and spills
Chemical Identifiers
| EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2) | |
| Does Not Contain Recycled Content | |
| Perforated Absorbent Pad | |
| 19 in. | |
| 15 in. | |
| 15 lb. |
| Black/Red on Yellow | |
| Polypropylene | |
| 19 x 15 in. (48.2 x 38 cm) | |
| 48.2 cm | |
| 38 cm | |
| 6.8 kg |
Specifications
| EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2) | |
| Does Not Contain Recycled Content | |
| 106 L | |
| Perforated Absorbent Pad | |
| Lightweight | |
| 19 in. | |
| 15 in. | |
| 15 lb. |
| Black/Red on Yellow | |
| 28 gal. | |
| Polypropylene | |
| 200/Cs. | |
| 19 x 15 in. (48.2 x 38 cm) | |
| 48.2 cm | |
| 38 cm | |
| 6.8 kg |