missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads

Catalog No. p-4492123
Change view
Click to view available options
Capacity (English):
22 gal.
28 gal.
Weight Capacity:
Heavyweight
Lightweight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Weight Capacity
296362243 22 gal. Heavyweight
19103101 28 gal. Lightweight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296362243 Supplier Brady™ Supplier No. CH100
Only null left

Color-coded absorbents for safe clean-up of hazardous fluids. Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads are surfactant treated polypropylene pads used on wide range of chemicals.

  • Benefits of sign and an absorbent in one
  • Highly absorbent for all-purpose industrial applications
  • Ideal for areas where the risk of slips, trips and falls are greatest
  • Chemically inert, do not react with aggressive fluids
  • Perforations provide cost control and reduce waste
  • Heavy weight - specifically designed for continuous leaks and larger spills
  • Light weight - specifically designed for smaller leaks, drips and spills

Specifications

Certifications/Compliance EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
Color Black/Red on Yellow
Description Does Not Contain Recycled Content
Capacity (English) 22 gal.
Capacity (Metric) 83.3 L
Material Polypropylene
Type Perforated Absorbent Pad
Packaging Quantity 100/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 19 x 15 in. (48.2 x 38 cm)
Length (English) 19 in.
Length (Metric) 48.2 cm
Width (English) 15 in.
Width (Metric) 38 cm
Weight (English) 15 lb.
Weight (Metric) 6.8 kg
Show More Show Less