missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Globally Harmonized System (GHS) Oxidizing Picto Labels

For oxidizers, organic peroxides

$10.58 - $13.88

Specifications

Certifications/Compliance GHS, OSHA 29 CFR 1910.1200
Color Black, Red, White
Format Label
Sterility Non-sterile
For Use With (Application) Oxidizers
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Material Size (English) Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Material Size (English) Size (Metric) Price Quantity & Availability  
19140514
View Documents Promotion Details
Brady™
118827
39/Pk. Vinyl 0.625 x 0.625 in. 1.58 x 1.58 cm
Pack of 39 Each for $13.88
Only null left
 
19140523
View Documents Promotion Details
Brady™
118881
39/Pk. Paper 0.625 x 0.625 in. 1.58 x 1.58 cm
Pack of 39 Each for $10.93
Only null left
 
19140532
View Documents Promotion Details
Brady™
118828
16/Pk. Vinyl 1 x 1 in. 2.54 x 2.54 cm
Pack of 16 Each for $12.74
Only null left
 
19140541
View Documents Promotion Details
Brady™
118882
16/Pk. Paper 1 x 1 in. 2.54 x 2.54 cm
Pack of 16 Each for $10.58
Only null left
 
19140701
View Documents Promotion Details
Brady™
118829
4/Card Vinyl (B-7569) 2 x 2 in. 5.08 x 5.08 cm
Pack of 4 Each for $11.89
Only null left
 
Description

Description

Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards

  • Legend: Oxidizing
  • Made of B-7569 Vinyl
  • Available in three sizes
Specifications

Specifications

GHS, OSHA 29 CFR 1910.1200
Label
Oxidizers
Black, Red, White
Non-sterile
Videos
SDS
Documents

Documents