missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Oxidizing Picto Labels
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Oxidizing
- Made of B-7569 Vinyl
- Available in three sizes
Specifications
Specifications
| Certifications/Compliance | GHS, OSHA 29 CFR 1910.1200 |
| Adhesive Type | Permanent Acrylic |
| Color | Black, Red, White |
| Finish | Gloss |
| Format | Label |
| Material | Vinyl (B-7569) |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Oxidizers |
| Quantity | 4/Card |
| Show More |