Learn More
Brady™ Globally Harmonized System (GHS) Oxidizing Picto Labels
For oxidizers, organic peroxides
Supplier:  Brady™ 118882
 View more versions of this product
Catalog No. 19140541
Only null left
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Oxidizing
  • Made of B-7569 Vinyl
  • Available in three sizes
Specifications
Chemical Identifiers
GHS, OSHA 29 CFR 1910.1200
Label
Oxidizers
Black, Red, White
Non-sterile
Specifications
GHS, OSHA 29 CFR 1910.1200
Black, Red, White
Label
Pictogram Label
Oxidizing
2 Years
1 x 1 in.
-40°F to 150°F
0.0038 in.
1 in.
2.5 cm
Acrylic, Pressure-sensitive
Semi-gloss
Paper
Non-sterile
Oxidizers
16/Pk.
2.54 x 2.54 cm
-40°C to +65°C
1 x 1 in.
1 in.
Product Suggestions
Product Suggestions