missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Globally Harmonized System (GHS) Environmental Picto Labels

For aquatic toxicity

$10.58 - $13.88

Specifications

Color Black, Red, White
Format Label
Sterility Non-sterile
For Use With (Application) For aquatic toxicity
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Material Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Material Price Quantity & Availability  
19140520
View Documents Promotion Details
Brady™
118863
39/Pk. Vinyl
Pack of 39 Each for $13.88
Only null left
 
19140529
View Documents Promotion Details
Brady™
118917
39/Pk. Paper
Pack of 39 Each for $10.58
Only null left
 
19140538
View Documents Promotion Details
Brady™
118864
16/Pk. Vinyl
Pack of 16 Each for $13.88
Only null left
 
19140547
View Documents Promotion Details
Brady™
118918
16/Pk. Paper
Pack of 16 Each for $11.68
Only null left
 
19140707
View Documents Promotion Details
Brady™
118865
4/Card Vinyl (B-7569)
Pack of 4 Each for $11.89
Only null left
 
Description

Description

Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards

  • Legend: Environmental
  • Made of B-7569 Viny or Paper
  • Available in three sizes
Specifications

Specifications

Black, Red, White
Label
Non-sterile
For aquatic toxicity
Videos
SDS
Documents

Documents