Learn More
Brady™ Globally Harmonized System (GHS) Environmental Picto Labels
For aquatic toxicity
Supplier:  Brady™ 118864
 View more versions of this product
Catalog No. 19140538
Only null left
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Environmental
  • Made of B-7569 Viny or Paper
  • Available in three sizes
Specifications
Chemical Identifiers
Black, Red, White
Label
Non-sterile
For aquatic toxicity
Specifications
GHS, OSHA 29 CFR 1910.1200
Black, Red, White
Label
Pictogram Label
Environmental
16/Pk.
-20°C to +75°C
1 x 1 in.
1 in.
Acrylic, Pressure-sensitive
Gloss
Vinyl
Non-sterile
For aquatic toxicity
-4°F to 167°F
0.004 in.
1 in.
2.5 cm
Product Suggestions
Product Suggestions