Learn More
Brady™ Globally Harmonized System (GHS) Environmental Picto Labels
For aquatic toxicity
Supplier:  Brady™ 118917
 View more versions of this product
Catalog No. 19140529
Only null left
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Environmental
  • Made of B-7569 Viny or Paper
  • Available in three sizes
Specifications
Chemical Identifiers
Black, Red, White
Label
Non-sterile
For aquatic toxicity
Specifications
GHS, OSHA 29 CFR 1910.1200
Black, Red, White
Label
Pictogram Label
Environmental
39/Pk.
-40°C to +65°C
0.625 x 0.625 in.
0.62 in.
Acrylic, Pressure-sensitive
Semi-gloss
Paper
Non-sterile
For aquatic toxicity
-40°F to 150°F
0.0038 in.
0.6 in.
15.20 mm
Product Suggestions
Product Suggestions