missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GHS Pictogram Signs

To identify equipment, rooms and materials, which contain or are contaminated with hazardous agents

$19.68 - $37.86

Specifications

Quantity 1 Ea.
Height (English) 14 in.
Height (Metric) 35.5 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19000478
View Documents Promotion Details
Brady™
133170
Polyester
Each for $19.68
Only null left
 
19000475
View Documents Promotion Details
Brady™
133167
Fiberglass
Each for $37.86
Only null left
 
Description

Description

  • Available in fiberglass and plastic material
  • Size: 35.5W x 25.4cm H(14 x 10 in.)

Specifications

Specifications

1 Ea.
35.5 cm
25.4 cm
GHS Pictogram Sign
14 in.
10 in.
GHS Pictograms
Videos
SDS
Documents

Documents