missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GHS Pictogram Signs

Catalog No. 19000478
Change view
Click to view available options
Material:
Fiberglass
Polyester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19000478 Polyester
19000475 Fiberglass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19000478 Supplier Brady™ Supplier No. 133170

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

To identify equipment, rooms and materials, which contain or are contaminated with hazardous agents

  • Available in fiberglass and plastic material
  • Size: 35.5W x 25.4cm H(14 x 10 in.)

Specifications

Material Polyester
Quantity 1 Ea.
Description Polyester (B-302)
Mounting Type Wall Mount
Height (English) 14 in.
Height (Metric) 35.5 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Shape Rectangular
Languages English
Dimensions (L x W) 25.4 x 35.5cm
Displayed Text GHS Pictograms
Product Type Safety Sign
Color Black, Red on White
Show More Show Less