missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GHS Pictogram Signs

Catalog No. 19000475
Change view
Click to view available options
Material:
Fiberglass
Polyester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19000475 Fiberglass
19000478 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19000475 Supplier Brady™ Supplier No. 133167
Only null left

To identify equipment, rooms and materials, which contain or are contaminated with hazardous agents

  • Available in fiberglass and plastic material
  • Size: 35.5W x 25.4cm H(14 x 10 in.)

Specifications

Material Fiberglass
Quantity 1 Ea.
Description Fiberglass (B-140)
Height (English) 14 in.
Height (Metric) 35.5 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Dimensions (L x W) 10 x 14 in.
Displayed Text GHS Pictograms
Product Type GHS Pictogram Sign