missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Danger: Hydrogen Flammable Gas No Smoking or Open Flames Signs
Used to identify equipment, rooms and materials which contain or are contaminated with hazardous agents
$13.62 - $47.33
Specifications
| Quantity | 1 Ea. |
|---|---|
| Min. Temperature (English) | -40°F |
| Displayed Text | DANGER Hydrogen Flammable Gas No Smoking or Open Flames |
| Product Type | Chemical and Hazardous Materials Sign |
| Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Thickness | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Thickness | Price | Quantity & Availability | ||
|
19035552
|
Brady™
69294 |
Fiberglass | 10 in. | 14 in. | 0.1 in. |
Each for $47.33
|
|
||
|
19036180
|
Brady™
25453 |
Plastic | 10 in. | 14 in. | 0.06 in. |
Each for $20.40
|
|
||
|
19036181
|
Brady™
84411 |
Polyester | 7 in. | 10 in. | 0.006 to 0.01 in. |
Each for $13.62
|
|
||
|
19036182
|
Brady™
84412 |
Polyester | 10 in. | 14 in. | 0.006 to 0.01 in. |
Each for $35.04
|
|
||
Description
- Available in variety of sizes and materials that include biohazard symbol
Specifications
| 1 Ea. | |
| -40°F | |
| DANGER Hydrogen Flammable Gas No Smoking or Open Flames | |
| Chemical and Hazardous Materials Sign |