missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Hydrogen Flammable Gas No Smoking or Open Flames Signs

Catalog No. 19036182
Change view
Click to view available options
Material:
Fiberglass
Plastic
Polyester
Height (English):
7 in.
10 in.
Width (English):
10 in.
14 in.
Thickness:
0.006 to 0.01 in.
0.06 in.
0.1 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English) Thickness
19036182 Polyester 10 in. 14 in. 0.006 to 0.01 in.
19035552 Fiberglass 10 in. 14 in. 0.1 in.
19036180 Plastic 10 in. 14 in. 0.06 in.
19036181 Polyester 7 in. 10 in. 0.006 to 0.01 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19036182 Supplier Brady™ Supplier No. 84412

Used to identify equipment, rooms and materials which contain or are contaminated with hazardous agents

  • Available in variety of sizes and materials that include biohazard symbol

Specifications

Material Polyester
Quantity 1 Ea.
Description Self-sticking Polyester
For Use With (Application) Safety sign, identification of fire alarm, extinguisher and emergency exit
Height (English) 10 in.
Height (Metric) 25.4 cm
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.006 to 0.01 in.
Width (English) 14 in.
Width (Metric) 35.5 cm
Temperature Range (Metric) -40°C to 110°C
Temperature Range (English) -40°F to 230°F
Displayed Text DANGER Hydrogen Flammable Gas No Smoking or Open Flames
Product Type Chemical and Hazardous Materials Sign
Color Red on White
Show More Show Less