missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Chemical and Hazardous Materials Signs: DANGER - EXPLOSIVE GAS NO SMOKING
Identify equipment, rooms and materials which contain or are contaminated with hazardous agents
$20.40 - $74.11
Specifications
| Certifications/Compliance | ANSI Z535, OSHA 1910.145 |
|---|---|
| Quantity | 1 Ea. |
| Mounting Type | Surface Mount |
| For Use With (Application) | Designed for use in industrial, utility, commercial and institutional environments |
| Min. Temperature (English) | -40°F |
| Catalog Number | Mfr. No. | Height (English) | Width (English) | Material | Thickness | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Height (English) | Width (English) | Material | Thickness | Price | Quantity & Availability | ||
|
19035940
|
Brady™
25651 |
10 in. | 14 in. | Plastic | 0.06 in. |
Each for $20.40
|
|
||
|
19035938
|
Brady™
72207 |
14 in. | 20 in. | Fiberglass | 0.1 in. |
Each for $74.11
|
|
||
Description
- Available in three materials - plastic, fiberglass and polyester
- Available in various widths and heights
- Black/red on white
Specifications
| ANSI Z535, OSHA 1910.145 | |
| Surface Mount | |
| -40°F | |
| Hazardous Materials Sign |
| 1 Ea. | |
| Designed for use in industrial, utility, commercial and institutional environments | |
| Explosive Gas No Smoking | |
| Red on White |