missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Chemical and Hazardous Materials Signs: DANGER - EXPLOSIVE GAS NO SMOKING

Catalog No. 19035940
Change view
Click to view available options
Height (English):
10 in.
14 in.
Width (English):
14 in.
20 in.
Material:
Fiberglass
Plastic
Thickness:
0.06 in.
0.1 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English) Material Thickness
19035940 10 in. 14 in. Plastic 0.06 in.
19035938 14 in. 20 in. Fiberglass 0.1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19035940 Supplier Brady™ Supplier No. 25651

Identify equipment, rooms and materials which contain or are contaminated with hazardous agents

  • Available in three materials - plastic, fiberglass and polyester
  • Available in various widths and heights
  • Black/red on white

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145
Material Plastic
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Height (English) 10 in.
Height (Metric) 25.40 cm
Max. Temperature (English) 175°F
Min. Temperature (English) -40°F
Thickness 0.06 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 80°C
Temperature Range (English) -40°F to 175°F
Displayed Text Explosive Gas No Smoking
Product Type Hazardous Materials Sign
Color Red on White
Show More Show Less