missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, 98%

CAS: 1303-86-2 | B2O3 | 69.62 g/mol

Supplier:  Thermo Scientific Chemicals A117070E

 View more versions of this product

Catalog No. AAA117070E


Only null left
Explore available promotions

Description

Description

Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Hygroscopic and moisture sensitive. Incompatible with acids and strong oxidizing agents.
Specifications

Specifications

1860°C
∽450°C
2500 g
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo(oxoboranyloxy)borane
518682
69.62
Odorless
2.46 +0.01
Boron oxide
1303-86-2
B2O3
14,1337
Soluble in water. Slightly soluble in methanol.
O=BOB=O
69.62
CHEBI:30163
99%
Hygroscopic

Chemical Identifiers

1303-86-2
69.62
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
oxo(oxoboranyloxy)borane
B2O3
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
CHEBI:30163
O=BOB=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360FD

EINECSNumber : 215-125-8

RTECSNumber : ED7900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only