missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, 98%

Catalog No. p-7081600
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1170736 500 g
AAA117070E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1170736 Supplier Thermo Scientific Chemicals Supplier No. A1170736
Only null left

CAS: 1303-86-2 | B2O3 | 69.62 g/mol

Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Hygroscopic and moisture sensitive. Incompatible with acids and strong oxidizing agents.

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo(oxoboranyloxy)borane
SMILES O=BOB=O

Specifications

Boiling Point 1860°C
Melting Point ∽450°C
Quantity 500 g
Merck Index 14,1337
Solubility Information Soluble in water. Slightly soluble in methanol.
Formula Weight 69.62
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.46 +0.01
Chemical Name or Material Boron oxide
Show More Show Less
Hazard Category H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 215-125-8
RTECSNumber ED7900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only