accessibility menu, dialog, popup

UserName

OBOBO

CAS RN 1303-86-2

IUPAC Name: oxo[(oxoboranyl)oxy]borane
Synonyms: boron oxide|boric oxide|boron trioxide|boric anhydride|diboron trioxide
Molecular Weight (g/mol): 69.62
Molecular Formula: B2O3
InChi Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N
SMILES: O=BOB=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (15)
Physical Form 
  • (5)
  • (3)
  • (7)
Percent Purity 
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (21)
  • (5)
  • (5)
  • (16)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (15)
  • (5)
  • (3)
  • (7)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)