missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron ICP Standard, 1000 ppm B in H2O/tr NH4OH, Ricca Chemical
$297.93 - $623.07
Chemical Identifiers
| CAS | 10043-35-3 |
|---|---|
| Molecular Formula | BH3O3 |
| Molecular Weight (g/mol) | 61.83 |
| MDL Number | MFCD00011337 |
| InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
| PubChem CID | 7628 |
| ChEBI | CHEBI:33118 |
| IUPAC Name | boric acid |
| SMILES | OB(O)O |
Chemical Identifiers
| 10043-35-3 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| CHEBI:33118 | |
| OB(O)O |
| BH3O3 | |
| MFCD00011337 | |
| 7628 | |
| boric acid |
Specifications
| Natural LDPE Bottle | |
| 97.0,0.56,0.1 | |
| Liquid | |
| MFCD00011337 | |
| 100°C | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 100 mL | |
| CHEBI:33118 | |
| Boron ICP Standard |
| 10043-35-3 , 7732-18-5 , 1336-21-6 | |
| 100.0,0.58,0.1 | |
| BH3O3 | |
| Colorless | |
| Standard | |
| OB(O)O | |
| 61.83 | |
| 7628 | |
| 1mL = 1mg B (1, 000ppm B)H3BO3 in H2O / trace NH4OH |
Safety and Handling
ShelfLife : 18 months