missing translation for 'onlineSavingsMsg'
Learn More

Boron ICP-MS Standard, 1000 ppm B in H2O/tr NH4OH, Ricca Chemical

Catalog No. MSB1KW100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSB1KW100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSB1KW100 Supplier Ricca Chemical Company Supplier No. MSB1KW100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10043-35-3 , 7732-18-5 , 1336-21-6
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm B
CAS Min % 97.0, 0.56, 0.1
CAS Max % 100.0, 0.58, 0.1
Equivalence 1 mL = 1 mg B
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Boron ICP-MS Standard
Show More Show Less