missing translation for 'onlineSavingsMsg'
Learn More

Boron AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg B), LabChem™

Catalog No. lc117801
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC117801 500 mL Poly Bottle
LC117807 125 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC117801 Supplier LabChem Supplier No. LC117801
Only null left

Chemical Identifiers

CAS 10043-35-3 , 7732-18-5
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Packaging Poly Bottle
CAS Max % 99.43, 0.57
Identification Passes Test
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg B)
Linear Formula H3BO3
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 61.83
Grade Certified
Chemical Name or Material Boron AA Standard, 1000ppm (1mL = 1mg B)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature