missing translation for 'onlineSavingsMsg'
Learn More

(-)-Bornyl acetate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

$259.32 - $759.05

Specifications

CAS 5655-61-8
Vapor Pressure 0.6 mmHg (20°C)
Density 0.986 g/mL (at 20°C)
Flash Point 84°C
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019039
View Documents
Merck Emd Millipore
458551MLF
1 mL
Each for $259.32
Only null left
 
111018232
View Documents
Merck Emd Millipore
458555MLF
5 mL
Each for $759.05
Only null left
 
Description

Description

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

5655-61-8
0.986 g/mL (at 20°C)
Neat
MFCD00867808
endo-(1 S)-1,7,7-Trimethylbicyclo[2.2.1 ]hept-2-yl acetate
196.29
Analytical Standard
0.6 mmHg (20°C)
84°C
C12H20O2
223°C to 224°C
Analytical Standard
≥99.0% (sum of enantiomers, GC)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'