missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Bornyl acetate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 458555MLF
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
| 5655-61-8 | |
| 0.986 g/mL (at 20°C) | |
| Neat | |
| MFCD00867808 | |
| endo-(1 S)-1,7,7-Trimethylbicyclo[2.2.1 ]hept-2-yl acetate | |
| 5 mL | |
| ≥99.0% (sum of enantiomers, GC) |
| 0.6 mmHg (20°C) | |
| 84°C | |
| C12H20O2 | |
| 223°C to 224°C | |
| Analytical Standard | |
| 196.29 | |
| Analytical Standard |