missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Certified, 4.0% (w/v) ±0.2%, LabChem™
$52.53 - $119.03
Chemical Identifiers
| CAS | 10043-35-3 |
|---|---|
| Molecular Formula | BH3O3 |
| Molecular Weight (g/mol) | 61.83 |
| MDL Number | MFCD00011337 |
| InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
| Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
| PubChem CID | 7628 |
| ChEBI | CHEBI:33118 |
| IUPAC Name | boric acid |
| SMILES | OB(O)O |
Chemical Identifiers
| 10043-35-3 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| 7628 | |
| boric acid |
| BH3O3 | |
| MFCD00011337 | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
| CHEBI:33118 | |
| OB(O)O |
Specifications
| Colorless | |
| 96,4 | |
| Poly Bottle | |
| Boron | |
| BH3O3 | |
| H3BO3 | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 7628 | |
| 61.83 | |
| Certified |
| 10043-35-3,7732-18-5 | |
| Passes Test | |
| Traceable to NIST | |
| 500 mL | |
| Liquid | |
| MFCD00011337 | |
| Soluble in water | |
| OB(O)O | |
| 61.83 | |
| CHEBI:33118 | |
| 4.0% ±0.2% (w/v) | |
| Boric Acid |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature