missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Certified, 4.0% (w/v) ±0.2%, LabChem™

Supplier:  LabChem LC117404

 View more versions of this product

Catalog No. LC117404

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Boric Acid
10043-35-3,7732-18-5
Passes Test
Traceable to NIST
4 L
Liquid
MFCD00011337
Soluble in water
OB(O)O
61.83
CHEBI:33118
4.0% ±0.2% (w/v)
Colorless
96,4
Poly Bottle
Boron
BH3O3
H3BO3
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83
Certified

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
OB(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

SDS
Documents

Documents

Promotions

Promotions