missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, ACS Reagent, Reagents
Boric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1104300500C2
Specifications
| Boric Acid, ACS Reagent | |
| 168°C to 170°C | |
| 300°C | |
| 1.435 | |
| BH3O3 | |
| MFCD00011337 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| ACS |
| Colorless/White | |
| 10043-35-3 | |
| Glass Bottle | |
| 500 g | |
| Solid | |
| Orthoboric Acid, Boracic Acid, Borofax, Trihydroxyborane | |
| OB(O)O | |
| 61.83 |
Chemical Identifiers
| 10043-35-3 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid |
| BH3O3 | |
| MFCD00011337 | |
| Orthoboric Acid, Boracic Acid, Borofax, Trihydroxyborane | |
| OB(O)O |