missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, ACS Reagent, Reagents

Catalog No. p-200021144
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338187 10 kg
01338186 3 kg
01338185 500 g
01338184 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01338187 Supplier Labchem Inc Supplier No. C110430010KG
Only null left

Boric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym Orthoboric Acid, Boracic Acid, Borofax, Trihydroxyborane
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Colorless/White
Melting Point 168°C to 170°C
Boiling Point 300°C
Packaging Poly Pail
Specific Gravity 1.435
Quantity 10 kg
Physical Form Solid
Grade ACS
Chemical Name or Material Boric Acid, ACS Reagent