missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, ACS Reagent, Reagents

Boric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc C110430050KG

 View more versions of this product

Catalog No. 01-338-184

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Boric Acid, ACS Reagent
168°C to 170°C
300°C
1.435
BH3O3
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
ACS
Colorless/White
10043-35-3
Fiber Drum
50 kg
Solid
Orthoboric Acid, Boracic Acid, Borofax, Trihydroxyborane
OB(O)O
61.83

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
BH3O3
MFCD00011337
Orthoboric Acid, Boracic Acid, Borofax, Trihydroxyborane
OB(O)O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions