missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 4% w/v, Reagents

Catalog No. 01335776
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335776 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335776 Supplier Labchem Inc Supplier No. CB11250020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Boric Acid, 4% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10043-35-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH 4.5 to 5
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.015
Quantity 20 L
Physical Form Liquid
Percent Purity 4%
Grade Reagent
Chemical Name or Material Boric Acid, 4% w/v