missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 4% (w/v) with Indicator (Bromocresol Green-Methyl Red), Certified, 4.0% (w/v) ±0.2%, LabChem™

Supplier:  LabChem LC117506

 View more versions of this product

Catalog No. LC117506


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Boric Acid
10043-35-3 , 7732-18-5 , 62625-32-5 , 845-10-3
BH3O3
MFCD00011337
Soluble in water
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83
Certified
Boron
10 L
4% (w/v) with Indicator (Bromocresol Green-Methyl Red)
4,95.998,0.001,0.001
H3BO3
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
Passes Test
OB(O)O
61.83
CHEBI:33118
4.0% ±0.2% (w/v)
Cubitainer™
Red
Liquid

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
CHEBI:33118
OB(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions