missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, 4% (w/v) with Indicator (Bromocresol Green-Methyl Red), Certified, 4.0% (w/v) ±0.2%, LabChem™
Supplier: LabChem LC117505
Specifications
| Boric Acid | |
| 10043-35-3 , 7732-18-5 , 62625-32-5 , 845-10-3 | |
| BH3O3 | |
| MFCD00011337 | |
| Soluble in water | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 7628 | |
| 61.83 | |
| Certified | |
| Boron | |
| 20 L |
| 4% (w/v) with Indicator (Bromocresol Green-Methyl Red) | |
| 4,95.998,0.001,0.001 | |
| H3BO3 | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe | |
| Passes Test | |
| OB(O)O | |
| 61.83 | |
| CHEBI:33118 | |
| 4.0% ±0.2% (w/v) | |
| Cubitainer™ | |
| Red | |
| Liquid |
Chemical Identifiers
| 10043-35-3 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| 7628 | |
| boric acid |
| BH3O3 | |
| MFCD00011337 | |
| orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe | |
| CHEBI:33118 | |
| OB(O)O |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature